Disodium-1,6-naphthalenedisulfonate 97%

Supplier: Ambeed
Warning

Synonyms: 1,6-Naphthalenedisulphonic acid Disodium salt, 1,6-Naphthalenedisulfonic acid Disodium salt, Sodium naphthalene-1,6-disulphonate, Sodium naphthalene-1,6-disulfonate

A529998-100G A529998-25G A529998-500G
76805-266EA 15 USD
76805-266 76805-268 76805-270
Disodium-1,6-naphthalenedisulfonate 97%
Disodium-1,6-naphthalenedisulfonate

Caution: for Research Use Only

Formula: C₁₀H₈O₆S₂
MW: 332.25 g/mol
MDL Number: MFCD00021522
CAS Number: 1655-43-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR