Dimidium bromide 98%

Supplier: Ambeed
Warning

A449281-100MG A449281-1G A449281-250MG A449281-5G
76804-498EA 21 USD
76804-498 76804-500 76804-502 76804-504
Dimidium bromide 98%
Dimidium bromide

Caution: for Research Use Only

Formula: C₂₀H₁₈BrN₃
MDL Number: MFCD00011757
CAS Number: 518-67-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR