4-Hydroxy-6-methylcoumarin 98%

Supplier: Ambeed
Warning

A431117-1G A431117-250MG A431117-5G
76804-282EA 30 USD
76804-282 76804-284 76804-286
4-Hydroxy-6-methylcoumarin 98%
4-Hydroxy-6-methylcoumarin

Caution: for Research Use Only

Formula: C₁₀H₈O₃
MDL Number: MFCD00085738
CAS Number: 13252-83-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR