Lithium dodecyl sulfate 98%

Supplier: Ambeed
Danger

A426512-5G
76804-216EA 5 USD
76804-216
Lithium dodecyl sulfate 98%
Lithium dodecyl sulphate

Caution: for Research Use Only

Formula: C₁₂H₂₅LiO₄S
MDL Number: MFCD00007467
CAS Number: 2044-56-6
UN: 1325
ADR: 4.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR