Isophthalonitrile 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1,3-Benzenedicarbonitrile, 1,3-Dicyanobenzene, Benzene-1,3-dicarbonitrile
A310400-100G
A310400-500G
A310400-25G
A310400-1000G
76802-748EA
15
USD
76802-748
76802-750
77306-022
77668-372
Isophthalonitrile 98%
Isophthalonitrile
Caution: for Research Use Only
Formula:
C₈H₄N₂ |
MDL Number:
MFCD00001795 CAS Number: 626-17-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...