EDTA (ethylenediamine tetraacetic acid) 98%
Supplier: Ambeed
|
Warning
|
A287527-100G
A287527-25G
A287527-500G
A287527-1000G
76802-326EA
9
USD
76802-326
76802-328
76802-330
77568-584
EDTA (ethylenediamine tetraacetic acid) 98%
EDTA (Ethylenediamine tetraacetic acid)
Caution: For research use only.
Formula:
C₁₀H₁₆N₂O₈ |
MDL Number:
MFCD00003541 CAS Number: 60-00-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...