4-[(4-Methyl-1-piperazinyl)sulfonyl]aniline 95%

Supplier: Ambeed
Warning

Synonyms: 4-((4-Methylpiperazin-1-yl)sulphonyl)aniline, 4-((4-Methylpiperazin-1-yl)sulfonyl)aniline

A277737-1G
76802-132EA 119 USD
76802-132
4-[(4-Methyl-1-piperazinyl)sulfonyl]aniline 95%
4-[(4-Methyl-1-piperazinyl)sulfonyl]aniline

Caution: for Research Use Only

Formula: C₁₁H₁₇N₃O₂S
MDL Number: MFCD03050283
CAS Number: 21623-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR