4-(4-Morpholinylcarbonyl)phenylboronic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(4-Morpholinylcarbonyl)benzeneboronic acid, (4-(Morpholine-4-carbonyl)phenyl)boronic acid
A256909-1G
A256909-250MG
A256909-5G
76801-702EA
63
USD
76801-702
76801-704
76801-706
4-(4-Morpholinylcarbonyl)phenylboronic acid 98%
4-(4-Morpholinylcarbonyl)phenylboronic acid
Caution: for Research Use Only
Formula:
C₁₁H₁₄BNO₄ |
MDL Number:
MFCD03411952 CAS Number: 389621-84-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...