Magnesium chloride hexahydrate 98%

Supplier: Ambeed
Warning

A252423-100G
76801-584EA 7 USD
76801-584 76801-586 76801-588
Magnesium chloride hexahydrate 98%
Magnesium chloride hexahydrate

Caution: for Research Use Only

Formula: MgCl₂·6H₂O
MDL Number: MFCD00149781
CAS Number: 7791-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR