Kojic acid 98%

Supplier: Ambeed
Warning

Synonyms: 5-Hydroxy-2-hydroxymethyl-4H-pyran-4-one, 5-Hydroxy-2-(hydroxymethyl)-4H-pyran-4-one, 5-Hydroxy-2-hydroxymethyl-4-pyrone, 5-Hydroxy-2-hydroxymethyl-γ-pyrone

A240690-100G A240690-25G A240690-25MG A240690-500G
103809-706EA 72 USD
103809-706 76801-314 76801-316 76801-318 77600-938
Kojic acid 98%
Kojic acid

Caution: for Research Use Only

Formula: C₆H₆O₄
Boiling Pt: 401.7 °C (760 mm Hg)
Melting Pt: 152…155 °C
Flash Pt: 179.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00006580
CAS Number: 501-30-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR