Magnesium trifluoromethanesulfonate 98%

Supplier: Ambeed
Danger

Synonyms: Magnesium trifluoromethanesulphonate, Magnesium triflate, Magnesium trifluoromethanesulfonate

A213421-25G A213421-5G
76800-698EA 23 USD
76800-698 76800-700
Magnesium trifluoromethanesulfonate 98%
Magnesium trifluoromethanesulphonate

Caution: for Research Use Only

Formula: C₂F₆MgO₆S₂
MDL Number: MFCD00042601
CAS Number: 60871-83-2
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR