Magnesium trifluoromethanesulfonate 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Magnesium trifluoromethanesulphonate, Magnesium triflate, Magnesium trifluoromethanesulfonate
A213421-25G
A213421-5G
76800-698EA
23
USD
76800-698
76800-700
Magnesium trifluoromethanesulfonate 98%
Magnesium trifluoromethanesulphonate
Caution: for Research Use Only
Formula:
C₂F₆MgO₆S₂ |
MDL Number:
MFCD00042601 CAS Number: 60871-83-2 UN: 3261 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...