DL-Malic acid 95%

Supplier: Ambeed
Warning

Synonyms: DL-Hydroxysuccinic acid, (±)-Malic acid, (±)-Hydroxy-1,4-butanedioic acid, (±)-Hydroxysuccinic acid, Hydroxybutanedioic acid, 2-Hydroxysuccinic acid

A200958-1000G A200958-100G A200958-25G A200958-500G
76800-346EA 13 USD
76800-346 76800-348 76800-350 76800-352
DL-Malic acid 95%
DL-Malic acid

Caution: for Research Use Only

Formula: HOOCCH(OH)CH₂COOH
MW: 134.09 g/mol
Boiling Pt: 306
MDL Number: MFCD00064212
CAS Number: 617-48-1
Merck Index: 13,05730

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR