JC-1 iodide salt 98%

Supplier: Ambeed
Warning

Synonyms: 5,5′,6,6′-Tetrachloro-1,1′,3,3′-tetraethylbenzimidazolocarbocyanine iodide, JC-1, 5,5′,6,6′-Tetrachloro-1,1′,3,3′-tetraethyl-imidacarbocyanine iodide, (E)-5,6-Dichloro-2-(3-(5,6-dichloro-1,3-diethyl-1,3-dihydro-2H-benzo[d]imidazol-2-ylidene)prop-1-en-1-yl)-1,3-diethyl-1H-benzo[d]imidazol-3-ium iodide, CBIC2(3), 1,1',3,3'-tetraethyl-5,5',6,6'-tetrachloroimidacarbocyanine iodide

A177382-1MG A177382-5MG
76799-566EA 101 USD
76799-566 76799-568
JC-1 iodide salt 98%
JC-1 iodide salt

Caution: for Research Use Only

Formula: C₂₅H₂₇Cl₄IN₄
MDL Number: MFCD00188475
CAS Number: 47729-63-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR