Ethyldiphenylphosphine 98%
Supplier: Ambeed
|
Warning
|
A162325-100G
A162325-1G
A162325-25G
A162325-5G
76799-050EA
173
USD
76799-050
76799-052
76799-054
76799-056
Ethyldiphenylphosphine 98%
Ethyldiphenylphosphine
Caution: for Research Use Only
Formula:
C₁₄H₁₅P |
MDL Number:
MFCD00015170 CAS Number: 607-01-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...