4-Chloro-6-(trifluoromethyl)quinoline 96%

Supplier: Ambeed
Warning

A151700-250MG A151700-25G A151700-5G A151700-1G
76798-668EA 17 USD
76798-668 76798-670 76798-672 77879-053
4-Chloro-6-(trifluoromethyl)quinoline 96%
4-Chloro-6-(trifluoromethyl)quinoline

Caution: for Research Use Only

Formula: C₁₀H₅ClF₃N
MDL Number: MFCD00153106
CAS Number: 49713-56-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR