4-Hydroxy-2-quinolinecarboxylic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Hydroxyquinoline-2-carboxylic Acid, Transtorine, Empirical Formula, Kynurenic acid, Quinurenic acid, KYNA, Kynuronic acid, Kinurenic acid
A141971-100MG
A141971-250MG
A141971-25MG
A141971-25G
A141971-5G
103803-084EA
32
USD
103803-084
76798-346
76798-348
76798-350
77316-286
77624-176
4-Hydroxy-2-quinolinecarboxylic acid 97%
4-Hydroxy-2-quinolinecarboxylic acid
Caution: for Research Use Only
Formula:
C₁₀H₇NO₃ Boiling Pt: 358.4 °C (760 mm Hg) Melting Pt: 275 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149476 CAS Number: 492-27-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...