D-(-)-Luciferin 98%

Supplier: Ambeed
Warning

Synonyms: D-Luciferin, (S)-2-(6-Hydroxybenzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylic acid

A122896-1MG A122896-1G A122896-250MG A122896-100MG A122896-5G
76797-482EA 6 USD
76797-482 77599-790 77599-792 77624-022 77878-906
D-(-)-Luciferin 98%
D(-)-Luciferin

Caution: for Research Use Only

Formula: C₁₁H₈N₂O₃S₂
MW: 280.3228 g/mol
MDL Number: MFCD00042929
CAS Number: 2591-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR