L-Leucine-tert-butyl ester hydrochloride 98%

Supplier: Ambeed
Warning

A122711-100G A122711-10G A122711-25G A122711-500G
103831-092EA 6 USD
103831-092 76797-460 76797-462 76797-464 77599-780
L-Leucine-tert-butyl ester hydrochloride 98%
L-Leucine-tert-butyl ester hydrochloride

Caution: for Research Use Only

Formula: C₁₀H₂₁NO₂
Flash Pt: 90.3 °C
Storage Temperature: Freezer
MDL Number: MFCD00038906
CAS Number: 2748-02-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR