L-Leucine-tert-butyl ester hydrochloride 98%
Supplier: Ambeed
|
Warning
|
A122711-100G
A122711-10G
A122711-25G
A122711-500G
103831-092EA
6
USD
103831-092
76797-460
76797-462
76797-464
77599-780
L-Leucine-tert-butyl ester hydrochloride 98%
L-Leucine-tert-butyl ester hydrochloride
Caution: for Research Use Only
Formula:
C₁₀H₂₁NO₂ Flash Pt: 90.3 °C Storage Temperature: Freezer |
MDL Number:
MFCD00038906 CAS Number: 2748-02-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...