VWR® One Spin Clinical Centrifuge
Supplier: VWR International
76795-570
76795-570EA 750.96 USD
76795-570
VWR® One Spin Clinical Centrifuge
Centrifuges Benchtop Centrifuges
The world’s best-selling clinical laboratory centrifuge designed to provide the ideal pre-analytic sample processing performance for all common blood tubes up to 75 to 100 mm (3 to 10 ml) with the press of just one button.
  • Preset runtime of 10 minutes at 1600×g
  • Simple two-button interface ensures repeatability and standardization, even across multiple sites
  • Process common all blood tubes 75 to 100 mm (3 to 10 ml) with no additional accessories
  • Swing-out horizontal rotor with 6 tube capacity
  • Clear, shatterproof locks during operation and will not open until the centrifuge has stopped
  • Cool-Flow air flow design prevents overheating of samples
  • 2 year warranty (lifetime on rotor)
Meet the VWR® One Spin Clinical Centrifuge, the world’s best-selling clinical laboratory centrifuge. Spin all common blood tubes up to 75 to 100 mm (3 to 10 ml) with the press of just one button. A preset speed and runtime (10 minutes at 1,600×g) make the VWR® One Spin Clinical Centrifuge the ideal choice for consistency in doctor’s offices, patient service centers, and outreach locations. Max speed 1600×g/3400 rpm.
Certifications: CE, MET, UKCA.
Delivery information: Supplied with horizontal rotor and tube holders.