VWR® One Spin Clinical Centrifuge with Timer
Supplier: VWR International
76795-568
76795-568EA 666.4 USD
76795-568
VWR® One Spin Clinical Centrifuge with Timer
Centrifuges Benchtop Centrifuges
Offers single-speed processing, simple controls, and an adjustable mechanical timer to meet a variety of blood specimen preparation needs.
  • Single g-force setting of 1600×g
  • Mechanical timer can be set from 1 to 30 min
  • Max speed 1600×g/3400 rpm
  • 6 tube capacity (75 to 100 mm/0.5 to 10 ml)
  • Small footprint, perfect for laboratories where space is at a premium
  • Cool-Flow air flow design prevents overheating of samples
  • 2 year warranty (lifetime on rotor)
Meet the One Spin Clinical Centrifuge with Timer, VWR®'s most affordable horizontal centrifuge. The One Spin Clinical Centrifuge with Timer offers single-speed processing, simple controls, and an adjustable mechanical timer to meet a variety of blood specimen preparation needs. Set to run at a constant 1,600×g, users can easily adjust cycle time from 1 to 30 minutes. Max speed 1600×g/3400 rpm.
Certifications: CE, MET, UKCA.
Delivery information: Supplied with horizontal rotor and universal tube holders.