Sterilization Wraps, 2-Ply SMS, One-Step Super Duty
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
KCRP1-600-0808 KCRP1-600-0909 KCRP1-600-0912 KCRP1-600-1212 KCRP1-600-1515 KCRP1-600-1818 KCRP1-600-1822 KCRP1-600-2020 KCRP1-600-2424 KCRP1-600-3030 KCRP1-600-3636 KCRP1-600-4040 KCRP1-600-4545 KCRP1-600-4548 KCRP1-600-4848 KCRP1-600-5454 KCRP1-600-5472 KCRP1-600-5490 KCRP1-600-6060 KCRP1-600-6080
76795-500CS 412.5 USD
76795-500 76795-530 76795-506 76795-532 76795-504 76795-538 76795-526 76795-522 76795-528 76795-534 76795-518 76795-508 76795-502 76795-536 76795-524 76795-510 76795-514 76795-512 76795-516 76795-520
Sterilization Wraps, 2-Ply SMS, One-Step Super Duty
Sterilization Wraps
One-Step heavy duty SMS 2-ply wrappers provide an excellent microbial barrier. Ideal for pharmaceutical production applications.
  • Manufactured and packaged in a certified ISO 8 Cleanroom
  • 2-ply
  • Clean - no cellulose
  • Strong, rip and tear resistant 50 gsm SMS material
  • An extensive range of sizes available means less cutting
  • Hydrophobic barrier allows sterilization gases, but prevents liquid and microbial penetration
  • Gram weight: 88 (each layer)
For use in steam, ethylene oxide and hydrogen peroxide sterilization processes.
Delivery information: Certificate of Conformance sent with each shipment.
Packaging: Sub packed into sealed labeled poly bags with lot number and expiration date for complete traceability.