Oakton Replacement Turbidity Calibration Set, Antylia Scientific
Supplier: Antyila Scientific
35635-52
76795-394EA 576.61 USD
76795-394
Oakton Replacement Turbidity Calibration Set, Antylia Scientific
Standards Turbidity Standards
For calibration of Oakton T-100 turbidity meters
  • Includes one each of 60-mL HPDE bottle of four NTU standards