Tin (II) sulfate 99%

Supplier: Ambeed
Danger

A957271-25G
76792-234EA 7 USD
76792-234
Tin (II) sulfate 99%
Tin (II) sulphate

Caution: for Research Use Only

Formula: O₄SSn
MDL Number: MFCD00011246
CAS Number: 7488-55-3
UN: 3260
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR