Trimethyl orthobutyrate 98% (by GC)

Supplier: Ambeed
Danger

Synonyms: Trimethylorthobutyrate, 1,1,1-Trimethoxybutane, Orthobutyric Acid Trimethyl Ester

A660790-25G
76790-890EA 40 USD
76790-890
Trimethyl orthobutyrate 98% (by GC)
Trimethyl orthobutyrate

Caution: for Research Use Only

Formula: C₇H₁₆O₃
MW: 148.20 g/mol
MDL Number: MFCD00008480
CAS Number: 43083-12-1
UN: 3272
ADR: 3 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR