Tin(II) chloride dihydrate 99%

Supplier: Ambeed
Danger

Synonyms: Stannous chloride dihydrate

A648741-25G
76790-852EA 18 USD
76790-852
Tin(II) chloride dihydrate 99%
Tin (II) chloride dihydrate

Caution: for Research Use Only

Formula: SnCl₂·2H₂O
MDL Number: MFCD00149863
CAS Number: 10025-69-1
UN: 3260
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR