2-Ethynyltoluene 95%

Supplier: Ambeed
Danger

Synonyms: 1-Ethynyl-2-methylbenzene, 2-Methylphenylacetylene

A621728-1G A621728-250MG A621728-5G
76790-664EA 13 USD
76790-664 76790-666 76790-668
2-Ethynyltoluene 95%
2-Ethynyltoluene

Caution: for Research Use Only

Formula: C₉H₈
MW: 116.1598 g/mol
MDL Number: MFCD03427202
CAS Number: 766-47-2
UN: 3295
ADR: 3 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR