Anthralin 98%

Supplier: Ambeed
Warning

Synonyms: 1,8,9-Trihydroxyanthracene, 1,8,9-Anthracenetriol, DIT, Dithranol, Anthranol, 1,8-Dihydroxyanthracen-9(10H)-one

A585109-1MG
76790-434EA 5 USD
76790-434
Anthralin 98%
Anthralin

Caution: for Research Use Only

Formula: C₁₄H₁₀O₃
MDL Number: MFCD00053409
CAS Number: 1143-38-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR