Thiazole 98%

Supplier: Ambeed
Danger

A510405-10G A510405-1G A510405-25G A510405-5G
76790-010EA 21 USD
76790-010 76790-012 76790-014 76790-016
Thiazole 98%

Caution: for Research Use Only

Formula: C₃H₃NS
MDL Number: MFCD00005315
CAS Number: 288-47-1
UN: 2924
ADR: 3,8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR