Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')aluminium 98%

Supplier: Ambeed
Warning

Synonyms: Al(TMHD)3, Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)aluminum

A471644-1G A471644-25G A471644-5G
76789-572EA 28 USD
76789-572 76789-574 76789-576
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')aluminium 98%
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')aluminium

Caution: for Research Use Only

Formula: C₃₃H₅₇AlO₆
MDL Number: MFCD00058918
CAS Number: 14319-08-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR