Tin(II) oxalate 99%

Supplier: Ambeed
Danger

Synonyms: Tin (II) ethanedioate, Oxalic acid tin (II) salt

A461111-100G A461111-500G A461111-25G
76789-480EA 26 USD
76789-480 76789-482 77669-072
Tin(II) oxalate 99%
Tin (II) oxalate

Caution: for Research Use Only

Formula: C₂O₄Sn
MW: 206.73 g/mol
MDL Number: MFCD00040678
CAS Number: 814-94-8
UN: 1759
ADR: 8 Ⅲ
Merck Index: 13,08865

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR