Tetraoctylammonium bromide 98%

Supplier: Ambeed
Warning

Synonyms: Tetra-n-octylammonium bromide

A408773-100G A408773-10G A408773-500G A408773-25G
103838-238EA 5 USD
103838-238 76789-010 76789-012 76789-014 76841-750
Tetraoctylammonium bromide 98%
Tetraoctylammonium bromide

Caution: for Research Use Only

Formula: C₃₂H₆₈BrN
Melting Pt: 95…98 °C
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00011863
CAS Number: 14866-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR