1-(2-Pyridylazo)-2-naphthol (PAN) 90% (by HPLC), Analytical reagent (AR)

Supplier: Ambeed
Warning

Synonyms: PAN, 1-(2-Pyridylazo)-2-naphthol

A402444-100G A402444-25G A402444-5G
76788-938EA 99 USD
76788-938 76788-940 76788-942
1-(2-Pyridylazo)-2-naphthol (PAN) 90% (by HPLC), Analytical reagent (AR)
1-(2-Pyridylazo)-2-naphthol

Caution: for Research Use Only

Formula: C₁₅H₁₁N₃O
MDL Number: MFCD00004071
CAS Number: 85-85-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR