1-(2-Pyridylazo)-2-naphthol (PAN) 90% (by HPLC), Analytical reagent (AR)
Supplier: Ambeed
|
Warning
|
Synonyms:
PAN, 1-(2-Pyridylazo)-2-naphthol
A402444-100G
A402444-25G
A402444-5G
76788-938EA
99
USD
76788-938
76788-940
76788-942
1-(2-Pyridylazo)-2-naphthol (PAN) 90% (by HPLC), Analytical reagent (AR)
1-(2-Pyridylazo)-2-naphthol
Caution: for Research Use Only
Formula:
C₁₅H₁₁N₃O |
MDL Number:
MFCD00004071 CAS Number: 85-85-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...