n-Tetradecane 99%

Supplier: Ambeed
Danger

A293460-25G
76787-594EA 6 USD
76787-594
n-Tetradecane 99%
n-Tetradecane

Caution: for Research Use Only

Formula: H₃C(CH₂)₁₂CH₃
MDL Number: MFCD00008986
CAS Number: 629-59-4
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR