SB202190 98%

Supplier: Ambeed
Warning

A277214-10MG A277214-1MG A277214-25MG A277214-50MG A277214-5MG A277214-100MG A277214-250MG
76787-328EA 45 USD
76787-328 76787-330 76787-332 76787-334 76787-336 77601-224 77872-336
SB202190 98%
SB202190

Caution: for Research Use Only

Formula: C₂₀H₁₄FN₃O
MDL Number: MFCD00941964
CAS Number: 152121-30-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR