Succinimide 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Pyrrolidine-2,5-dione
A168239-1000G
A168239-100G
A168239-25G
A168239-500G
76784-130EA
27
USD
76784-130
76784-132
76784-134
76784-136
Succinimide 97%
Succinimide
Caution: for Research Use Only
Formula:
C₄H₅NO₂ MW: 99.09 g/mol Boiling Pt: 285 |
MDL Number:
MFCD00005495 CAS Number: 123-56-8 Merck Index: 12,09040 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...