ZSTK474 ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4,4'-(6-(2-(Difluoromethyl)-1H-benzo[d]imidazol-1-yl)-1,3,5-triazine-2,4-diyl)dimorpholine
A156873-10MG
A156873-1MG
A156873-50MG
76783-638EA
20
USD
76783-638
76783-640
76783-642
ZSTK474 ≥98%
ZSTK474
Caution: for Research Use Only
Formula:
C₁₉H₂₁F₂N₇O₂ |
MDL Number:
MFCD08705334 CAS Number: 475110-96-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...