2,6-Dichloro-3-cyanopyrazine 95%

Supplier: Ambeed
Warning

A156868-100G A156868-10G A156868-250MG A156868-25G A156868-5G A156868-1G
103804-928EA 22 USD
103804-928 103843-132 76783-632 76783-634 76783-636 77300-220 77300-222 77318-938
2,6-Dichloro-3-cyanopyrazine 95%
2,6-Dichloro-3-cyanopyrazine

Caution: for Research Use Only

Formula: C₅HCl₂N₃
Boiling Pt: 276.6 °C (760 mm Hg)
Flash Pt: 121 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13193460
CAS Number: 313339-92-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR