tert-Butyl-4-fluorobenzoate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl 4-fluorobenzoate
A156240-100G
A156240-10G
A156240-1G
A156240-25G
A156240-500G
A156240-5G
76783-582EA
79
USD
76783-582
76783-584
76783-586
76783-588
76783-590
76783-592
tert-Butyl-4-fluorobenzoate 98%
tert-Butyl 4-fluorobenzoate
Caution: for Research Use Only
Formula:
C₁₁H₁₃FO₂ |
MDL Number:
MFCD01320759 CAS Number: 58656-98-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...