tert-Butyl-4-fluorobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4-fluorobenzoate

A156240-100G A156240-10G A156240-1G A156240-25G A156240-500G A156240-5G
76783-582EA 79 USD
76783-582 76783-584 76783-586 76783-588 76783-590 76783-592
tert-Butyl-4-fluorobenzoate 98%
tert-Butyl 4-fluorobenzoate

Caution: for Research Use Only

Formula: C₁₁H₁₃FO₂
MDL Number: MFCD01320759
CAS Number: 58656-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR