Triethylammonium bromide 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Triethylamine hydrobromide
A146386-100G
A146386-10G
A146386-25G
76783-166EA
43
USD
76783-166
76783-168
77317-076
Triethylammonium bromide 95%
Triethylammonium bromide
Caution: for Research Use Only
Formula:
C₆H₁₅N·HBr MW: 182.1 |
MDL Number:
MFCD00054287 CAS Number: 636-70-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...