Vandetanib 98%
Supplier: Ambeed
|
Warning
|
A126372-1MG
A126372-25MG
A126372-5MG
A126372-250MG
A126372-25G
76782-038EA
5
USD
76782-038
76782-040
76782-042
77871-573
77871-574
Vandetanib 98%
Vandetanib
Caution: for Research Use Only
Formula:
C₂₂H₂₄BrFN₄O₂ |
MDL Number:
MFCD07772346 CAS Number: 443913-73-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...