Replacement Air Filter for ULT Freezers
Supplier: Thermo Fisher Scientific
332297H02
76779-128EA 172.66 USD
76779-128
Replacement Air Filter for ULT Freezers
Freezers Ultra Low Temperature Freezers
Replacement air filter for ultra-low temperature chest freezers.
Routine maintenance is critical for proper ultra-low temperature (ULT) freezer operation. Replacing your ULT's old air filter enables the necessary airflow to the unit's refrigeration system.