Portelite™ Fluorimetric Total Nucleic Acid Quantitation Kit
Supplier: AAT BIOQUEST INC
17631
76779-116EA 332.82 USD
76779-116
Portelite™ Fluorimetric Total Nucleic Acid Quantitation Kit
Assays Nucleic Acid Assays
Portelite™ fluorimetric total nucleic acid quantitation kit is optimized for measuring the total amounts of nucleic acids with CytoCite™ or Qubit® fluorometers.
  • Optimized for cytocite and qubit fluorometers
  • Sensitive and indiscriminate binding
The kit has all the essential reagents, including Helixyte™ Green ssDNA reagent, dilution buffer, and pre-diluted DNA standards.
Packaging: Plastic vial.
Caution: For research use only.