ProLite™ His-Tag Protein Gel Staining Kit, AAT Bioquest
Supplier: AAT BIOQUEST INC
18010
76779-082EA 444.33 USD
76779-082
ProLite™ His-Tag Protein Gel Staining Kit, AAT Bioquest
Protein Expression Protein Expression Reagents
This kit requires very little hands-on time, allowing for rapid protein expression screening in a variety of gel types using an imaging equipment with a standard FITC filter.
  • Green fluorescence
  • Compatible with standard FITC filter
Kit provides a fast, sensitive, and highly specific fluorescent stain for visualizing His-tagged fusion proteins directly in a polyacrylamide gel following electrophoresis
It detects nanogram of His-tagged proteins directly in gels, thus eliminating the additional western blotting step.
Caution: For research use only.