ReadiLink™ iFluor® 488 FISH Fluorescence Imaging Kit
Supplier: AAT Bioquest
17310
76779-026EA 534.09 USD
76779-026
ReadiLink™ iFluor® 488 FISH Fluorescence Imaging Kit
Protein Linking and Labeling Systems
The kit provides Taq DNA polymerase enzyme, which incorporates iFluor® 488-dUTPs in the target DNA through Polymerase Chain Reaction (PCR).
  • Robust protocol
  • Bright labeling
  • Photostable fluorescence
The Readilink™ iFluor® 488 FISH fluorescence imaging kit is a convenient tool for labeling a target DNA using an iFluor® 488 labeled FISH probe via in situ hybridization.
Our proprietary iFluor® dyes are brighter and more photostable than traditional fluorescent labels, providing the desired resolution and signal.
Packaging: Packaged in plastic vials.
Caution: For research use only.