Linearity FLQ Fertility for Siemens Atellica/Centaur, AUDIT MircoControls
Supplier: AUDIT MICROCONTROLS
K818M-5
76778-984EA 546.16 USD
76778-984
Linearity FLQ Fertility for Siemens Atellica/Centaur, AUDIT MircoControls
Controls and Calibration Verification Materials
The Audit® MicroControlsTM Linearity FLQ Fertility for Siemens Atellica/Centaur is intended to simulate human patient samples for use in determining linearity, calibration verification, and the verification of reportable range for the following analytes: AFP, DHEA-S, Estradiol, Progesterone, FSH, HCG, LH, Prolactin, Testosterone.
  • Frozen Liquid Format
Consists of 10 levels of liquid material and additives in bovine serum albumin.
Caution: For In Vitro Diagnostic use only.