INDICAID® COVID-19 OTC Rapid Tests 2/PK FDA EUA for OTC Use, for Persons Aged 2 Years or Older
Supplier: Germaine Laboratories
P0038
76778-982CS 957.6 USD
76778-982 76778-983
INDICAID® COVID-19 OTC Rapid Tests 2/PK FDA EUA for OTC Use, for Persons Aged 2 Years or Older
Clinical Diagnostic Supplies Clinical Reagents and Kits
INDICAID® COVID-19 OTC RAPID TEST 2/PK. A lateral flow immunoassay intended for the qualitative detection of SARS-CoV-2. FDA EUA Authorized for Over-the-counter use, for persons aged two years or older. Minimum order quantity applies.
  • A lateral flow immunoassay intended for the qualitative detection of SARS-CoV-2
  • FDA EUA Authorized for Over-the-counter use
  • For persons aged two years or older
  • Minimum order quantity applies
The INDICAID® COVID-19 Rapid Antigen At-Home Test is a lateral flow immunoassay intended for the qualitative detection of SARS-CoV-2. This test is authorized for non-prescription home use with self-collected direct anterior Nasal (nares) swab samples from individuals 14 years or older with symptoms of COVID-19 within the first six days of symptom onset.
This test is also intended for non-prescription home use with adult-collected anterior nasal (nares) swab samples from individuals aged two years or older with symptoms of COVID-19 within the first 6 days of symptom onset.
Certifications: FDA EUA for OTC.
Ordering information: Lead time applies.